Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134812.1 | NP_001128284.1 | 2 - 3 | 882 | 77 | |
GenBank mRNA | BC169182.1 | - | 2 - 3 | 882 | 77 | |
CT030607.1 | - | 2 - 3 | 862 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |