Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134813.2 | NP_001128285.1 | 3 - 4 | 449 | 85 | |
XM_012965835.2 | XP_012821289.1 | 3 - 4 | 715 | 85 | ||
XM_012965836.2 | XP_012821290.1 | 3 - 4 | 675 | 85 | ||
XM_012965837.2 | XP_012821291.1 | 3 - 4 | 419 | 85 | ||
XM_012965838.2 | XP_012821292.1 | 3 - 4 | 346 | 85 | ||
GenBank mRNA | CT030404.1 | - | 3 - 4 | 431 | 85 | |
CX962582.1 | - | 3 - 4 | 449 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |