Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137611.1 | NP_001131083.1 | 2 - 3 | 222 | 89 | |
GenBank mRNA | EF564347.1 | - | 2 - 3 | 222 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |