Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142012.1 | NP_001135484.1 | 2 - 3 | 236 | 80 | |
GenBank mRNA | BC167695.1 | - | 2 - 3 | 236 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |