Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142044.1 | NP_001135516.1 | 8 - 9 | 1241 | 74 | |
XM_012956554.1 | XP_012812008.1 | 10 - 11 | 1235 | 74 | ||
XM_012956555.1 | XP_012812009.1 | 9 - 10 | 1154 | 74 | ||
XM_012956556.1 | XP_012812010.1 | 9 - 10 | 1372 | 74 | ||
XM_012956557.1 | XP_012812011.1 | 9 - 10 | 1000 | 74 | ||
GenBank mRNA | BC167935.2 | - | 8 - 9 | 1241 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |