Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142064.1 | NP_001135536.1 | 2 - 3 | 797 | 97 | |
XM_012968107.2 | XP_012823561.1 | 2 - 3 | 889 | 97 | ||
GenBank mRNA | BC168005.1 | - | 2 - 3 | 797 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |