Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142068.1 | NP_001135540.1 | 4 - 5 | 1030 | 69 | |
XM_012966199.2 | XP_012821653.1 | 4 - 5 | 1297 | 69 | ||
XM_012966200.2 | XP_012821654.1 | 4 - 5 | 1298 | 69 | ||
XM_012966201.2 | XP_012821655.1 | 4 - 5 | 1298 | 69 | ||
XM_012966202.2 | XP_012821656.1 | 4 - 5 | 1298 | 69 | ||
XM_012966203.2 | XP_012821657.1 | 4 - 5 | 1297 | 69 | ||
XM_012966204.2 | XP_012821658.1 | 4 - 5 | 1297 | 69 | ||
XM_012966206.2 | XP_012821660.1 | 4 - 5 | 1296 | 69 | ||
XM_012966207.2 | XP_012821661.1 | 4 - 5 | 1296 | 69 | ||
XM_012966208.2 | XP_012821662.1 | 4 - 5 | 1295 | 69 | ||
XM_012966209.2 | XP_012821663.1 | 4 - 5 | 1295 | 69 | ||
XM_012966210.2 | XP_012821664.1 | 4 - 5 | 1296 | 69 | ||
XM_018095295.1 | XP_017950784.1 | 4 - 5 | 1297 | 69 | ||
GenBank mRNA | BC168010.1 | - | 4 - 5 | 1030 | 69 | |
CT030480.1 | - | 2 - 3 | 685 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |