Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkdcc.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142110.1 | NP_001135582.2 | 1 - 2 | 542 | 78 | |
XM_018096176.1 | XP_017951665.1 | 1 - 2 | 1354 | 78 | ||
GenBank mRNA | BC168090.1 | - | 1 - 2 | 542 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |