Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mkrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142112.1 | NP_001135584.1 | 4 - 5 | 521 | 105 | |
GenBank mRNA | BC168097.1 | - | 4 - 5 | 521 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |