Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142119.1 | NP_001135591.1 | 3 - 4 | 642 | 58 | |
GenBank mRNA | BC168116.1 | - | 3 - 4 | 642 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |