Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142155.1 | NP_001135627.1 | 2 - 3 | 427 | 93 | |
XM_012956575.2 | XP_012812029.1 | 2 - 3 | 413 | 93 | ||
GenBank mRNA | BC168479.1 | - | 2 - 3 | 427 | 93 | |
CT025515.2 | - | 2 - 3 | 274 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |