Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142195.1 | NP_001135667.1 | 4 - 5 | 1385 | 93 | |
GenBank mRNA | BC168576.1 | - | 4 - 5 | 1385 | 93 | |
CT027881.2 | - | 3 - 4 | 1292 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |