Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grpel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142216.1 | NP_001135688.1 | 1 - 2 | 89 | 96 | |
GenBank mRNA | BC168632.1 | - | 1 - 2 | 89 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |