Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142224.1 | NP_001135696.1 | 3 - 4 | 303 | 62 | |
XM_012964399.2 | XP_012819853.1 | 4 - 5 | 476 | 62 | ||
XM_012964400.2 | XP_012819854.1 | 3 - 4 | 404 | 62 | ||
GenBank mRNA | BC168804.1 | - | 3 - 4 | 303 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |