Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142224.1 | NP_001135696.1 | 9 - 10 | 1027 | 114 | |
XM_012964399.2 | XP_012819853.1 | 10 - 11 | 1200 | 114 | ||
XM_012964400.2 | XP_012819854.1 | 9 - 10 | 1128 | 114 | ||
GenBank mRNA | BC168804.1 | - | 9 - 10 | 1027 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |