Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142909.1 | NP_001136381.1 | 5 - 6 | 1001 | 94 | |
XM_012966096.2 | XP_012821550.1 | 6 - 7 | 1521 | 94 | ||
GenBank mRNA | BC161420.1 | - | 5 - 6 | 1001 | 94 | |
CR926386.2 | - | 6 - 7 | 1085 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |