Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145748.1 | NP_001139220.1 | 4 - 5 | 341 | 73 | |
GenBank mRNA | BC077053.1 | - | 4 - 5 | 334 | 73 | |
BC161642.1 | - | 4 - 5 | 318 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |