Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hbz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005092.2 | NP_001005092.1 | 3 | 448 | 114 | |
GenBank mRNA | BC077020.1 | - | 3 | 449 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |