Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spaca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011312.1 | NP_001011312.1 | 4 | 1050 | 93 | |
GenBank mRNA | BC088040.1 | - | 4 | 1050 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |