Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ras-dva1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011503.2 | NP_001011503.2 | 1 | 751 | 101 | |
GenBank mRNA | AY729885.1 | - | 1 | 817 | 101 | |
BC170576.1 | - | 1 | 722 | 101 | ||
BC170578.1 | - | 1 | 722 | 101 | ||
CR942596.2 | - | 1 | 751 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |