Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hist1h4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016869.2 | NP_001016869.1 | 1 | 245 | 102 | |
GenBank mRNA | CR848633.2 | - | 1 | 245 | 102 |
Gene Symbol: LOC100493978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018097408.1 | XP_017952897.1 | 1 | 653 | 102 |
Gene Symbol: LOC100497406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018091714.1 | XP_017947203.1 | 2 | 1082 | 102 |
Gene Symbol: LOC101732670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004913817.3 | XP_004913874.2 | 1 | 325 | 102 |
Gene Symbol: LOC101733933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018097175.1 | XP_017952664.1 | 1 | 695 | 102 |
Gene Symbol: LOC101735268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018090569.1 | XP_017946058.1 | 1 | 229 | 102 |
Gene Symbol: LOC105947139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012962051.2 | XP_012817505.1 | 1 | 436 | 102 |
Gene Symbol: LOC105947140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012962052.2 | XP_012817506.1 | 1 | 1257 | 102 |
Gene Symbol: LOC108645245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018090216.1 | XP_017945705.1 | 1 | 250 | 102 |
Gene Symbol: LOC108648868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018097426.1 | XP_017952915.1 | 1 | 507 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Xenopus tropicalis Both primers and probe map within a single exon |