Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dlx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030395.1 | NP_001025566.1 | 650 | 104 | ||
GenBank mRNA | BC090605.1 | - | 650 | 104 | ||
CR942714.2 | - | 676 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |