Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ces2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078719.1 | NP_001072187.1 | 13 | 2055 | 86 | |
XM_012961287.2 | XP_012816741.1 | 14 | 2112 | 86 | ||
GenBank mRNA | BC082503.1 | - | 13 | 2055 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |