Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100207.1 | NP_001093677.1 | 2 | 1050 | 96 | |
XM_012959745.2 | XP_012815199.1 | 2 | 1812 | 96 | ||
GenBank mRNA | BC135589.1 | - | 2 | 1050 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |