Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsf6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127160.1 | NP_001120632.1 | 452 | 61 | ||
GenBank mRNA | BC161785.1 | - | 452 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |