Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144014.1 | NP_001137486.1 | 1259 | 87 | ||
GenBank mRNA | AY603492.1 | - | 1259 | 87 | ||
AY639018.1 | - | 1339 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Zea mays |