Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100283810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156708.1 | NP_001150180.1 | 331 | 81 | ||
GenBank mRNA | EU966026.1 | - | 331 | 81 |