Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158243.1 | NP_001151715.1 | 171 | 60 | ||
GenBank mRNA | BT042218.1 | - | 171 | 60 | ||
EU971983.1 | - | 230 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |