Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111831.1 | NP_001105301.1 | 1171 | 78 | ||
GenBank mRNA | AY106054.1 | - | 1186 | 78 | ||
D49475.1 | - | 1171 | 78 | |||
U93560.1 | - | 1051 | 78 | |||
U93561.1 | - | 1051 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Zea mays |