Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPP2-B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147195.1 | NP_001140667.1 | 343 | 81 | ||
GenBank mRNA | BT039323.1 | - | 343 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Zea mays |