Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152929.1 | NP_001146401.1 | 332 | 69 | ||
GenBank mRNA | BT055116.1 | - | 332 | 69 | ||
BT055282.1 | - | 329 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |