Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100284444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157339.1 | NP_001150811.1 | 345 | 91 | ||
GenBank mRNA | BT042024.1 | - | 243 | 91 | ||
EU958859.1 | - | 251 | 91 | |||
EU968394.1 | - | 345 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |