Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111877.1 | NP_001105347.1 | 972 | 84 | ||
GenBank mRNA | AY111861.1 | - | 972 | 84 | ||
BT053782.1 | - | 938 | 84 | |||
EU966040.1 | - | 988 | 84 | |||
M74566.1 | - | 972 | 84 |