Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ypt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112076.1 | NP_001105546.1 | 706 | 80 | ||
GenBank mRNA | X63277.1 | - | 706 | 80 |