Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100285161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158055.1 | NP_001151527.1 | 1310 | 100 | ||
GenBank mRNA | EU971062.1 | - | 1310 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |