Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001148123.1 | NP_001141595.1 | 965 | 84 | ||
GenBank mRNA | BT041686.1 | - | 965 | 84 | ||
BT063423.1 | - | 636 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |