Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100281015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153934.1 | NP_001147406.1 | 454 | 122 | ||
GenBank mRNA | EU955272.1 | - | 454 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Zea mays |