Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100281351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154269.1 | NP_001147741.1 | 237 | 70 | ||
GenBank mRNA | BT062039.1 | - | 198 | 70 | ||
EU956416.1 | - | 237 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |