Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147614.1 | NP_001141086.1 | 346 | 72 | ||
GenBank mRNA | BT040366.1 | - | 346 | 72 | ||
BT085766.1 | - | 312 | 72 | |||
BT088241.1 | - | 226 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Zea mays |