Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100283515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156415.1 | NP_001149887.1 | 303 | 71 | ||
GenBank mRNA | EU964988.1 | - | 303 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |