Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100284943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157838.1 | NP_001151310.1 | 254 | 56 | ||
GenBank mRNA | EU970224.1 | - | 254 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |