Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111451.1 | NP_001104921.1 | 61 | 85 | ||
GenBank mRNA | AB564720.1 | - | 41 | 85 | ||
AF084478.3 | - | 61 | 85 | |||
AY110044.1 | - | 61 | 85 | |||
EU959731.1 | - | 85 | 85 |