Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111548.1 | NP_001105018.1 | 891 | 73 | ||
GenBank mRNA | AF318580.1 | - | 891 | 73 |