Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111549.1 | NP_001105019.1 | 1835 | 84 | ||
GenBank mRNA | AF322918.1 | - | 1835 | 84 | ||
BT033627.1 | - | 1872 | 84 |