Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111726.1 | NP_001105196.1 | 220 | 68 | ||
GenBank mRNA | AY156978.1 | - | 220 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |