Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111529.1 | NP_001104999.1 | 170 | 57 | ||
GenBank mRNA | AF256229.1 | - | 170 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |