Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111698.1 | NP_001105168.1 | 685 | 87 | ||
GenBank mRNA | AY029313.1 | - | 685 | 87 | ||
AY105309.1 | - | 685 | 87 | |||
BT063312.1 | - | 716 | 87 |