Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111806.1 | NP_001105276.1 | 441 | 88 | ||
GenBank mRNA | AY738149.1 | - | 441 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Zea mays |