Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111901.1 | NP_001105371.1 | 1179 | 93 | ||
GenBank mRNA | AY103866.1 | - | 1316 | 93 | ||
U17973.1 | - | 1204 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |