Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111933.1 | NP_001105403.1 | 985 | 91 | ||
GenBank mRNA | AY106556.1 | - | 1053 | 91 | ||
BT068528.1 | - | 1078 | 91 | |||
EU968473.1 | - | 1079 | 91 | |||
U83625.1 | - | 985 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Zea mays |